Seventh Generation® Free and Clear Natural 2X Concentrate Laundry Detergent
500044-327
: Avantor
- Size:150 oz.
- Environmentally Preferable:Sustainable Materials
- Environmental Certification:USDA Certified Biobased - 97%
- Chemical/physical specifications:Plant-derived surfactants
- Cat. No.:500044-327
- Packaging:Bottle
- Supplier no.:SEV22803
Concentrated formula means more loads washed per bottle than regular liquid detergents
- Unscented Liquid
- Washes 99 loads
- No optical brighteners
- Not tested on animals
The combination of plant-based substances and enzymes gets out the toughest stains, grease and grime. Nontoxic, biodegradable, hypoallergenic, and free of chlorine, phosphates and petroleum-based ingredients. No artificial fragrances or dyes. Works in high-efficiency or standard machines.