ITW Dymon Stainless Steel Cleaner Towels
Catalog # 500037-786
Supplier: Janitorial Supplies
Specifications
- Width:9¾"
- Description:Premoistened Cloth, Lemon scented, 11/2 lbs.
- Length:10½"
- Cat. No.:500037-786
- Packaging:Canister
- Supplier no.:ITW91930
- No. of towels/wipes per container:30
Specifications
About this item
Stainless steel cleaner and polish in convenient premoistened towels.
- D-Limonene and Mineral oil based
- Removes fingerprint, grease and water mark
Clean, polish, brighten and protect in one step without dripping or overspraying. Prevent water marks and corrosion while preserving the high luster of stainless steel. For use on stainless, steel, chrome and aluminum.
Certifications: NSF Registered.