ContactUsLinkComponent
Universal® Heavy-Duty Stapler
Universal® Heavy-Duty Stapler
Catalog # 500025-580
undefined
Universal® Heavy-Duty Stapler
Catalog # 500025-580
Supplier Number:  UNV43048
Specifications
  • Sheet capacity:
    200
  • Color:
    Black/Graphite
  • Description:
    Rear load heavy-duty stapler
  • Material:
    Metal/plastic
  • Cat. No.:
    500025-580
  • Supplier no.:
    UNV43048
Specifications
About this item
Powerhouse drives through up to 200 sheets with a single staple
  • Features extra throat depth and adjustable paper stopper for precision stapling
  • Nonskid base and grip handle offer secure, comfortable operation
  • Durable metal and plastic construction
  • Rear loads heavy-duty staples
Replacement staples: SWI35320