ContactUsLinkComponent
Swingline® Heavy-Duty Long Reach™ Stapler
Swingline® Heavy-Duty Long Reach™ Stapler
Catalog # 500025-587
undefined
Swingline® Heavy-Duty Long Reach™ Stapler
Catalog # 500025-587
Supplier Number:  SWI34121
Specifications
  • Sheet capacity:
    20
  • Color:
    Black
  • Description:
    Heavy-duty stapler for standard staples
  • Material:
    Metal
  • Cat. No.:
    500025-587
  • Supplier no.:
    SWI34121
Specifications
About this item
Feature-packed stapler allows you to staple up to 12" from the paper's edge—perfect for center-stitch stapling or fastening cards or tags
  • Integrated ruler and adjustable locking paper guide ensure precision alignments and consistency
  • Rugged metal construction
  • Mounts to table or bench
  • Easy-open, drop-in channel loading of a full strip (210) of standard staples
Uses standard 0.25" staples.
16 3/4" length, 12" throat depth.