Order Entry
United States
ContactUsLinkComponent
Rubbermaid® Commercial Square Brute® Container
Rubbermaid® Commercial Square Brute® Container
Catalog # 500006-470
undefined
Rubbermaid® Commercial Square Brute® Container
Catalog # 500006-470
Supplier Number:  RCP352600GY

Specifications

  • Depth:
    21½"
  • Ø:
    21 1/2"
  • Width:
    21½"
  • Capacity:
    28 gal.
  • Color:
    Gray
  • Description:
    Trash container with side handles, top opening
  • Height:
    22½"
  • Material:
    Plastic
  • Cat. No.:
    500006-470
  • Shape:
    Square
  • For use with:
    Rubbermaid® Commercial Square Brute® lid RUB352700GY
  • Supplier no.:
    RCP352600GY

Specifications

About this item

Holds 14% more than round containers in same space.

Molded handles and bottom grip.