Pendaflex® Portable Letter Size File Box
Catalog # 500010-008
Supplier: Janitorial Supplies
Specifications
- Color:Black
- Dimensions:147/8×113/4×111/4" (Outer); 137/8×103/4×101/4" (Inner)
- Material:Plastic
- Size:Letter
- Cat. No.:500010-008
- For use with:Hanging Files
- Supplier no.:ESS20861
Specifications
About this item
If your workload is increasing, here's just the thing to keep things properly sorted.
Holds up to 20 letter-size, 1/2" capacity hanging folders (not included). Modular design with flip-down handle makes for easy stacking. With integrated dual side grips and latch-locking lid.It has dual side grip and flip down handle.