Order Entry
United States
ContactUsLinkComponent
Advantus® Panel Wall Wire Hooks, Essendant LLC MS
Advantus® Panel Wall Wire Hooks, Essendant LLC MS
  500013-524
undefined
Advantus® Panel Wall Wire Hooks, Essendant LLC MS
  500013-524
 :  AVT75370

 

  • Color:
    Silver
  • Description:
    Panel wall wire hooks, plastic backing
  • Height:
    2½"
  • Environmentally Preferable:
  • Cat. No.:
    500013-524
  • Depth:
    0.90"
  • Width:
    3.40"
  • Supplier no.:
    AVT75370

 

 

Keep lightweight objects on the panel wall and off desk, freeing valuable desk space.

  • Metal wire hangers hold lightweight objects
  • Panel accessory type: Hook

Hang pictures, calendars or keys-hooks easily insert into fabric panel walls. Wire hangers also clip notes or memos between hanger and panel.