Order Entry
United States
ContactUsLinkComponent
Dataproducts® R0167 Cash Register Ribbon, Essendant
Dataproducts® R0167 Cash Register Ribbon, Essendant
Catalog # 500024-004
undefined
Dataproducts® R0167 Cash Register Ribbon, Essendant
Catalog # 500024-004
Supplier Number:  DPSR0167

Specifications

  • Color:
    Black, Red
  • Description:
    Cash register ribbon, nylon, compatible
  • Cat. No.:
    500024-004
  • Supplier no.:
    DPSR0167

Specifications

About this item

For use with Citizen® DP-600 Series, LDP-3530, LDP-3535 and LDP-3540 cash registers

Consistent, crisp character output.