Lexmark™ 1040990, 1040993 Printer Ribbon
Catalog # 500036-304
Supplier: Janitorial Supplies
Specifications
- For Use With:IBM® 6400 , 6408 printers
- Color:Black
- Description:OEM nylon printer ribbon
- Environmentally Preferable:
- Cat. No.:500036-304
- Supplier no.:LEX1040990
Specifications
About this item
20,000,000 character yield.