Order Entry
United States
ContactUsLinkComponent
Acroprint® Model ATR120 Electronic Time Clock Ribbon, Essendant
Acroprint® Model ATR120 Electronic Time Clock Ribbon, Essendant
Catalog # 500023-963
undefined
Acroprint® Model ATR120 Electronic Time Clock Ribbon, Essendant
Catalog # 500023-963
Supplier Number:  ACP390127000

Specifications

  • Color:
    Blue/red
  • Description:
    OEM ribbon for time clock ATR120
  • Material:
    Polyester/plastic
  • Cat. No.:
    500023-963
  • Supplier no.:
    ACP390127000

Specifications

About this item

For use with Acroprint® ATR120 weekly/biweekly payroll clock

Accurate, clear output.