Acroprint® Model 125 Heavy-Duty Manual Print Time Recorder Ribbon, Essendant
500024-040
- Color:Blue/red
- Description:OEM ribbon for time clock
- Material:Polyester/plastic
- Cat. No.:500024-040
- Supplier no.:ACP200106002
For use with Acroprint® manual print time recorders models 125 and 150
Accurate, clear output.