Order Entry
United States
ContactUsLinkComponent
Rolodex™ Mesh Stacking Sorter
Rolodex™ Mesh Stacking Sorter
Catalog # 500025-105
undefined
Rolodex™ Mesh Stacking Sorter
Catalog # 500025-105
Supplier Number:  ROL22141

Specifications

  • Depth:
    14¼"
  • Width:
    8½"
  • Color:
    Black
  • Description:
    Inclined file sorter
  • Height:
    7½"
  • Material:
    Mesh Metal
  • Cat. No.:
    500025-105
  • For use with:
    Stacking Side Load Letter Tray (ROL22321 Series)
  • No. of compartments:
    5
  • Supplier no.:
    ROL22141

Specifications

About this item

Contemporary mesh stacking sorter adds style, efficiency and productivity to your desktop

Helps you make the most of your valuable desktop space. Provides five sections for file folders, books, notepads, binders and more. Stacks on top of Stacking Side Load Letter Tray, sold separately.