Order Entry
United States
ContactUsLinkComponent
Acroprint® ES1000 Electronic Totalizing Payroll Recorder Time Clock Ribbon, Essendant
Acroprint® ES1000 Electronic Totalizing Payroll Recorder Time Clock Ribbon, Essendant
Catalog # 500023-917
undefined
Acroprint® ES1000 Electronic Totalizing Payroll Recorder Time Clock Ribbon, Essendant
Catalog # 500023-917
Supplier Number:  ACP390129000

Specifications

  • Color:
    Black/red
  • Description:
    OEM ribbon for 1000ES time clock
  • Material:
    Polyester/plastic
  • Cat. No.:
    500023-917
  • Supplier no.:
    ACP390129000

Specifications

About this item

For use with Acroprint® electronic payroll recorder model ES1000

Accurate, clear output.