HON® Core Removable Lock Kit, Essendant LLC MS
Catalog # 500016-811
Supplier: Janitorial Supplies
Specifications
- For Use With:HON® Credenzas, HON® Desks, HON® Lateral Files, HON® Workstations
- Color:Black
- Cat. No.:500016-811
- Supplier no.:HONF23BX
Specifications
About this item
Lock with a removable cylinder that can be interchanged as needed.
Allows users to access all furniture with a single key for convenience and security. Makes re-keying quick and easy. Order one core per unit.