Order Entry
United States
Orders LinkContactUsLinkComponent
Matrix Printer Labels, Dots, Bulk Pack, Universal

Matrix Printer Labels, Dots, Bulk Pack, Universal

Supplier:   Janitorial Supplies
Matrix Printer Labels, Dots, Bulk Pack, Universal
Certificates
Matrix Printer Labels, Dots, Bulk Pack, Universal
Matrix Printer Labels, Dots, Bulk Pack, Universal

1 Options Available Below

Product Details & Documents

Print labels for mass mailings from your pin-fed printer.

  • Perforated at the page fold after every 12" across one label
  • Rectangular shape

Self-adhesive labels apply quickly.

Product Family Options

Product Information

  • Material
    Color
    For Use With
    Dimensions
    Availability
    Price
  • 500016-220
    Matrix printer labels, dots, bulk pack
    Matrix printer labels, dots, bulk pack
    Paper
    White
    Dot matrix printers
    1.44×4"
    Discontinued
    Supplier Number: -
  • 500016-212
    Matrix printer labels, dots, bulk pack
    Matrix printer labels, dots, bulk pack
    Paper
    White
    Dot matrix printers
    3.5×0.44"
    Discontinued
    Supplier Number: -
  • 500016-208
    Matrix printer labels, dots, bulk pack
    Matrix printer labels, dots, bulk pack
    Paper
    White
    Dot matrix printers
    0.94×3.5"
    Log in for availability
    Log in for availability
    Box of 5,000
    Box of 5,000 - $27.71
    $27.71
    Box of 5,000
       $27.71
    Minimum order quantity is 1

    Supplier Number: UNV70104
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.