Order Entry
United States
ContactUsLinkComponent
Dial® Boraxo® Original Powdered Hand Soap
Dial® Boraxo® Original Powdered Hand Soap
Catalog # 500036-602
Supplier:  Avantor
undefined
Dial® Boraxo® Original Powdered Hand Soap
Catalog # 500036-602
Supplier:  Avantor

Specifications

  • Capacity:
    5 lbs.
  • Color:
    White
  • Description:
    Hand soap, unscented
  • Cat. No.:
    500036-602
  • Supplier no.:
    DIA02203EA

Specifications

About this item

Gently scrub away the deepest grime.

  • Contains no phosphates
  • 100% soluble

It will not clog drains or harm fixtures. Gentle on hands due to the select blend of quality soap and borax.