Epson® ERC23B, ERC23BR Cash Register Ribbon, Essendant
Catalog # 500024-123
Supplier: Janitorial Supplies
Specifications
- Color:Black, Red
- Description:Cash register ribbon, nylon, OEM
- Cat. No.:500024-123
- Supplier no.:EPSERC23BR
Specifications
About this item
For use with Epson® 250, 252, 255, 257, 260, 262, 265, 267, RP-265, RP-267, TM-267 and TM-300 cash registers
Long life and exceptional character definition.