Lathem® Time VIS6008 Electronic Time Recorder Replacement Ribbon, Essendant
Catalog # 500023-898
Supplier: Janitorial Supplies
Specifications
- Color:Purple
- Description:OEM ribbon for 1000E time clock
- Cat. No.:500023-898
- Supplier no.:LTHVIS6008
Specifications
About this item
For use with Latham 900E, 1000E, 1500E, 5000EP, 7000E and 7500E electronic time recorders
Accurate, clear output.