House of Doolittle™ Weekly Appointment Book Ruled without Appointment Times, Essendant
500003-882
- Paper color:White
- Sheet size:6⅞×8¾"
- Environmentally Preferable:Sustainable Materials
- Cover Color:Black
- Calendar Term:12 months (Jan.-Dec.)
- Separate Sections:Notes, telephone/address
- Binding Type:Wire
- Calendar Format:Current week with 4 months
- Cover Material:Simulated leather
- Cat. No.:500003-882
- Supplier no.:HOD25802
- Page format:1 week per 2-page spread
Ruled for writing in appointment times for a flexible schedule.
- Columnar format provides one column per weekday and one for Saturday and Sunday combined
- Extras include telephone/address and memo sections
Seasonal availability.