Order Entry
United States
ContactUsLinkComponent
HON® Removable Lock Core Replacement Kit, Essendant LLC MS
HON® Removable Lock Core Replacement Kit, Essendant LLC MS
  500016-810
   Sustainable Materials
   Safer Human and Environmental Health
undefined
HON® Removable Lock Core Replacement Kit, Essendant LLC MS
  500016-810
 :  HONF23CX
   Sustainable Materials
   Safer Human and Environmental Health

 

  • For Use With:
    HON® Products
  • Color:
    Brushed Chrome
  • Material:
    Metal, Brushed Chrome
  • Environmentally Preferable:
    Sustainable Materials,
    Safer Human and Environmental Health
  • Environmental Certification:
    level® 2 Certified, Scientific Certification Systems (SCS) Certified Indoor Advantage™ Gold
  • Cat. No.:
    500016-810
  • Supplier no.:
    HONF23CX

 

 

For files that have HON® “One Key” standard locks.

Replaces the lock core, allowing all files to be keyed alike. Order one like-numbered kit per file.

 : Scientific Certification Systems (SCS) Certified Indoor Advantage™ Gold.