Dataproducts® E2276 Cash Register Ribbon, Essendant
Catalog # 500023-865
Supplier: Janitorial Supplies
Specifications
- Color:Purple
- Description:Cash register ribbon, nylon, compatible, dot matrix
- Cat. No.:500023-865
- Supplier no.:DPSE2276
Specifications
About this item
For use with Epson® ERC-35, 875 and M875 cash registers
Consistent, trouble-free performance.