Dataproducts® R2116 Cash Register Ribbon, Essendant
Catalog # 500023-878
Supplier: Janitorial Supplies
Specifications
- Color:Purple
- Description:Cash register ribbon, fabric, compatible
- Cat. No.:500023-878
- Supplier no.:DPSR2116
Specifications
About this item
For use with Epson® TM-U200D, 300A, 300B, 300C, 300D, 370 and 375 cash registers
Consistent, trouble-free performance.