Dataproducts® P6010 Printer Ribbon, Essendant
Catalog # 500024-052
Supplier: Janitorial Supplies
Specifications
- For Use With:Oki® Microline , Okimate printers
- Color:Black
- Description:Compatible nylon printer ribbon
- Cat. No.:500024-052
- Supplier no.:DPSP6010
Specifications
About this item
This printer Ribbon has consistent and trouble-free performance.
- Used for dot matrix printer