Dataproducts® R5190 Printer Ribbon, Essendant
Catalog # 500023-920
Supplier: Janitorial Supplies
Specifications
- For Use With:IBM® , pPS™ printers
- Color:Black
- Description:Compatible nylon printer ribbon
- Environmentally Preferable:
- Cat. No.:500023-920
- Supplier no.:DPSR5190
Specifications
About this item
This printer Ribbon has long life and exceptional character definition.
- Used for dot matrix printer
- Re-Inkable ribbon