Order Entry
United States
Orders LinkContactUsLinkComponent
Pendaflex® SureHook™ Nine-Section Hanging File Folder

Pendaflex® SureHook™ Nine-Section Hanging File Folder

Classification hanging file folder, Blue, Letter, ⅓, 3½"
Catalog # 500011-861
Marketsource
   Sustainable Materials
Supplier:   Janitorial Supplies
Supplier #:  PFX59225
Pendaflex® SureHook™ Nine-Section Hanging File Folder
Catalog # 500011-861
Marketsource
   Sustainable Materials
Pendaflex® SureHook™ Nine-Section Hanging File Folder
Pendaflex® SureHook™ Nine-Section Hanging File Folder

Classification hanging file folder, Blue, Letter, ⅓, 3½"

Price
$38.84
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Features SureHook™ technology with revolutionary built in tension springs.

  • With 9 Sections

Rods and hooks are stronger and sturdier with less bending. Folders stay on rails and slide with ease. Preprinted indexed A to Z tabs in assorted positions. Rip-proof tape reinforced gusset tops. Includes 10 self-adhesive blank labels.

Certifications: Acid Free, SFI Certified.

Specifications
  • Color:
    Blue
  • Size:
    Letter
  • Type:
    Classification hanging file folder
  • Expansion:
    3½"
  • Tab cut:
  • Cat. No.:
    500011-861
  • Environmental Certification:
    Sustainable Forestry Initiative® (SFI) Certified
  • Environmentally Preferable:
    Sustainable Materials
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.