TallyGenicom® 1A3066B01 Printer Ribbon, Essendant
Catalog # 500023-999
Supplier: Janitorial Supplies
Specifications
- For Use With:Texas Instruments 8920 , 8930 printers
- Color:Black
- Description:OEM nylon printer ribbon
- Environmentally Preferable:
- Cat. No.:500023-999
- Supplier no.:TEX1A3066B01
Specifications
About this item
This has precision pairing between printer and ribbon for easy installation and trouble-free use.
- Has 15000000 character yield
- Used for dot matrix printer