Dataproducts® R8600 Matrix Printer Ribbon, Essendant
Catalog # 500023-942
Supplier: Janitorial Supplies
Specifications
- For Use With:Basic Four/MAI, Data Royal, Datasouth, Infoscribe , printek® printers
- Color:Black
- Description:Compatible nylon printer ribbon
- Cat. No.:500023-942
- Supplier no.:DPSR8600
Specifications
About this item
This matrix printer ribbon has consistent, crisp character output.
- Used for dot matrix printer