Dataproducts® R6405 Printer Ribbon, Essendant
Catalog # 500023-935
Supplier: Janitorial Supplies
Specifications
- For Use With:Panasonic® printers
- Color:Black
- Description:Compatible nylon printer ribbon
- Cat. No.:500023-935
- Supplier no.:DPSR6405
Specifications
About this item
This printer ribbon has long life and exceptional character definition.
- Used for dot matrix printer