LYSOL® Brand I.C.™ Quaternary Disinfectant Cleaner
Catalog # 500033-170
Supplier: Avantor
Specifications
- Description:Liquid Disinfetant Cleaner, Natural scented
- Size:1 gal.
- Environmentally Preferable:
- Cat. No.:500033-170
- Supplier no.:RAC74983CT
Specifications
About this item
Hospital-strength multipurpose disinfectant cleans, disinfects and deodorizes hard, nonporous surfaces.
- Neutral pH
- Ammonia based
- Economical, concentrated formula makes up to 256 gallons
Virucidal, fungicidal and bactericidal. Effective against Pseudomonas, Salmonella, Staph, MRSA, Strep, E.coli, and HIV-1 (AIDS), herpes simplex 1 and 2, RSV, influenza A2 (H1N1), and Hepatitis B viruses, among others, including animal viruses.
Certifications: EPA Registered.