C-Line® Looseleaf CD/DVD Organizer Sheet
Catalog # 500024-674
Supplier: Janitorial Supplies
Specifications
- For Use With:CD Holder CLI61955, Three-Ring Binder
- Capacity:4
- Color:Clear/White
- Cat. No.:500024-674
- Supplier no.:CLI61958
Specifications
About this item
Double-sided sheets hold CDs and DVDs.
- Media sleeve features: Liner
Punched to fit a standard three-ring binder or CD case (sold separately). Made of polypropylene, so the discs easily slide in and out.