Quartet® Flexible Metal Cubicle Panel Hangers
Catalog # 500013-127
Supplier: Janitorial Supplies
Specifications
- For Use With:1¹/₂ - 2¹/₂" thick partition walls
- Color:Silver
- Height:2¼"
- Environmentally Preferable:
- Cat. No.:500013-127
- Depth:2¼"
- Width:1¼"
- Supplier no.:QRT7501
Specifications
About this item
These strong partition hangers can make cubicle more efficient. Hang a whiteboard or bulletin board right in your workspace and can be more productive.
- Silver, nickel-plated finish hangers match board frame
- Two hangers per set
- Over the panel mounting
- Flexible metal material
Turn wasted space into a hard working communication center hang planning or message boards from cubicle wall panels 1.5 to 2.5" thick. Compatible for use with Oval Office™ fabric bulletin boards and most Quartet® aluminum frame boards.