Order Entry
United States
ContactUsLinkComponent
Rolodex™ Distinctions™ Business Card Holder, Essendant
Rolodex™ Distinctions™ Business Card Holder, Essendant
  500013-317
 :  Avantor
undefined
Rolodex™ Distinctions™ Business Card Holder, Essendant
  500013-317
 :  Avantor

 

  • For Use With:
    Cards size 2.25×4"
  • Capacity:
    50
  • Color:
    Metal/Black
  • Description:
    Business card holder
  • Dimensions:
    2×2.8×4.9"
  • Material:
    Wood/metal
  • Cat. No.:
    500013-317
  • Supplier no.:
    ROLE23578

 

 

Classic meets industrial in this smart combination of punched metal and wood

Open design accommodates business cards of almost any size. High back holds cards upright. Nonskid feet.