Order Entry
United States
ContactUsLinkComponent
 
Scotch® ATG 700 Adhesive Transfer Tape Applicator
  500000-058
undefined
Scotch® ATG 700 Adhesive Transfer Tape Applicator
  500000-058
 :  MMMATG700

 

  • For Use With:
    Tape width: 0.5, 0.75"; tape length: 1296"
  • Color:
    Yellow
  • Description:
    Handheld dispenser
  • Cat. No.:
    500000-058
  • Supplier no.:
    MMMATG700

 

 

Pull trigger and lay down adhesive

Great for wrapping, mounting, projects of all kinds. Covered gears minimize adhesive residue and applicator jams. For use with 1/2" and 3/4" wide rolls. Clear cover allows you to see when tape is running out. Use with Scotch® Adhesive Transfer Tape MMM9241/2 or MMM9243/4 (sold separately).