PM Company® SecurIT® 5" Self-Locking Plastic Seals
Catalog # 500024-533
Supplier: Avantor
Specifications
- Color:Clear
- Length:5"
- Material:Plastic
- Cat. No.:500024-533
- Supplier no.:PMC04636
Specifications
About this item
Designed to stay securely locked through shipment.
- Notched strip provides various locking positions
- Easily threads through and locks into place
- Cannot be pulled apart; removable only when cut
- Durable weatherproof plastic construction
For use with transit sacks, sold separately.