House of Doolittle™ Economy Daily Desk Calendar Refill, Essendant
500003-903
- Paper color:White
- Sheet size:3.5×6"
- Description:Calendar refill for use with #17 bookstyle base
- Environmentally Preferable:Sustainable Materials
- Appointment Ruling:7 AM to 5 PM, Mon.-Sat.
- Calendar Term:12 months (Jan.-Dec.)
- Calendar Format:Current day with current month, past month
- Cat. No.:500003-903
- Supplier no.:HOD4717
- Page format:1 page Saturday, 1 page Sunday, 1 weekday per two-page spread
One full page for appointments and one full page for notes each weekday.
- One page each for Saturday and Sunday appointments
- Reference calendars make scheduling easier
Seasonal availability.