ContactUsLinkComponent
C-Line® Traditional Sheet Protector, Essendant
C-Line® Traditional Sheet Protector, Essendant
Catalog # 500022-633
undefined
C-Line® Traditional Sheet Protector, Essendant
Catalog # 500022-633
Supplier Number:  CLI00010
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Color:
    Clear
  • Description:
    Heavy Grade Ring Binder Sheet Protector, Three Side Load
  • Material:
    PP
  • Cat. No.:
    500022-633
  • Insert size:
    11×8.5"
  • Supplier no.:
    CLI00010
  • No. of holes punched:
    3
Specifications
About this item
Three hole punch for 11 x 8 ½ punched sheets
Foldover on binding edge for reinforcement and securing of contents. Open on three sides. Polypropylene with black mounting sheets.