Sealed Air Bubble Wrap® AirCap® Air Cellular Cushioning Material
500018-080
- Description:Packaging with bubble cushioning type, with perforation for every 12"
- Length:100'
- Thickness:0.8 cm (0.3")
- Cat. No.:500018-080
- Width:12"
- Supplier no.:SEL48561
Strong-grade cushioning material is ideal for fragile products with long or unpredictable distribution cycles.
- Lightweight packaging reduces costs and provides superior protection
- Provides better protection using less packaging material compared to loose fill or paper
This packaging retains air and cushioning properties much longer than non-barrier bubbles.