Westcott® Transparent Plastic Ruler, Essendant
Metric/imperial ruler, Plastic, mm, cm; ¹⁄₁₆", 6"
Catalog # 500024-323
Supplier: Janitorial Supplies
Supplier #: ACM45016
Jump to:
Product Details & Documents
Transparent plastic construction
1" wide.
Specifications
- Description:Metric/imperial ruler
- Length:6"
- Material:Plastic
- Graduations:mm, cm; ¹⁄₁₆"
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



