3M™ Electronic Equipment Cleaning Wipes
Catalog # 500037-780
Supplier: Janitorial Supplies
Specifications
- Width:5½"
- Color:White
- Length:6¾"
- Cat. No.:500037-780
- Packaging:Canister
- Supplier no.:MMMCL610
- No. of towels/wipes per container:80
Specifications
About this item
Pre-moistened wipes keeps our equipment clean and dust-free.
- Non-abrasive and safe for use on most electronic surfaces
- Cuts through grime and grease
These antistatic wipes are safe to use on electronic equipment and delicate surfaces including LCD screens and DVD media. Wipes come in a canister, keeping them moist and allowing for quick and easy dispensing.