Order Entry
United States
ContactUsLinkComponent
Kantek LCD Monitor Stand
Kantek LCD Monitor Stand
Catalog # 500025-396
undefined
Kantek LCD Monitor Stand
Catalog # 500025-396
Supplier Number:  KTKMS200B

Specifications

  • W×D:
    18×12½"
  • Color:
    Black
  • Description:
    Monitor stand with two drawers
  • Height:
    5"
  • Cat. No.:
    500025-396
  • Supplier no.:
    KTKMS200B

Specifications

About this item

Designed to help reduce neck and eye strain by keeping monitor at a comfortable angle.

  • Constructed from high impact plastic to support LCDs up to 35 lbs.
  • Rubber feet prevent falling off of surface

Drawers provide ample storage space to keep office supplies organized and off the desk.