ContactUsLinkComponent
simple green® d Pro 5 One Step Disinfectant
simple green® d Pro 5 One Step Disinfectant
  500033-192
undefined
simple green® d Pro 5 One Step Disinfectant
  500033-192
 :  SMP30501
  • Description:
    Liquid disinfectant bottle, Unscented
  • Size:
    1 gal.
  • Cat. No.:
    500033-192
  • Supplier no.:
    SMP30501
All-in-one disinfectant, sanitizer and cleaner is highly effective against HIV-1 (AIDS virus), Hepatitis B and C, MRSA, Avian Influenza (bird flu), Athlete's foot fungus, Influenza A and many Gram-positive and Gram-negative bacteria. Prevents growth of mold and mildew.
  • Ammonia based
  • No added color or fragrance
Suitable for use in hospitals and food processing facilities. Applicble on ceramic, fiberglass, plastic, porcelain, stainless steel and tile.
Dilutes 1:16 for heavy cleaning, 1:64 for cleaning and disinfecting, and 1:128 for sanitizing.