AirguardĀ® Synthetic Pocket Filters, Total Filtration
Supplier: Total Filtration Services
Certificates
About this item
Extended surface pocket filters feature efficiency, high dust holding capacity, long service life, and low resistance to air flow.
Applications include general ventilation, stand-alone single-stage filtration, built-up frame banks, side-access systems, rooftop air handlers, air supply for paint spray booths, and economical upgrade from fiberglass or synthetic-framed disposable filters.
Ordering information: Extended surface pocket filters are available in ratings from MERV 9 to MERV 14; contact your Avantor representative for more information.
Specifications
- Catalog No:
- 78001-462
- 78001-530
- 78001-460
- 78001-538
- 78001-534
- 78001-544
- 78001-546
- 78001-540
- 78001-548
- 78001-470
- 78001-478
- 78001-472
- 78001-550
- 78001-552
- 78001-480
- 78001-482
- 78001-558
- 78001-556
- 78001-564
- 78001-498
- 78001-496
- 78001-506
- 78001-508
- 78001-574
- 78001-504
- 78001-570
- 78001-580
- 78001-576
- 78001-440
- 78001-438
- 78001-448
- 78001-444
- 78001-510
- 78001-512
- 78001-524
- 78001-526
- 78001-528
- 78001-454
- 78001-456
- 78001-450
- 78001-452
- Dimensions:
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 30.5 cm (24 x 12")
- 61 x 30.5 cm (24 x 12")
- 61 x 61 cm (24 x 24")
- 61 x 30.5 cm (24 x 12")
- 61 x 30.5 cm (24 x 12")
- 61 x 30.5 cm (24 x 12")
- 61 x 30.5 cm (24 x 12")
- 61 x 30.5 cm (24 x 12")
- 61 x 30.5 cm (24 x 12")
- 61 x 30.5 cm (24 x 12")
- 61 x 30.5 cm (24 x 12")
- 61 x 30.5 cm (24 x 12")
- 61 x 30.5 cm (24 x 12")
- 30.5 x 61 cm (12 x 24")
- 61 x 51 cm (24 x 20")
- 61 x 51 cm (24 x 20")
- 51 x 51 cm (20 x 20")
- 51 x 51 cm (20 x 20")
- 61 x 51 cm (24 x 20")
- 51 x 61 cm (20 x 24")
- 61 x 51 cm (24 x 20")
- 51 x 51 cm (20 x 20")
- 61 x 51 cm (24 x 20")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")