Megabase DNA Standards, Lonza
Catalog # 12002-108
Supplier: Avantor
CAS Number:
Specifications
- Concentration:10 ±2 µg DNA/plug
- Description:Lambda DNA Ladders
- Cat. No.:12002-108
- Range:50 - 873 kb
- Supplier no.:50401
Specifications
About this item
Megabase DNA standards are prepared in InCert® agarose gel plug format for easy handling in pulsed field gel electrophoresis
Each lot is performance tested for reliable results. Store at 2 to 8°C (36 to 46°F). All standards come with five plugs each