


Buffered oxide etch (6:1) VLSI, with surfactant, J.T.Baker®
JT5569-3
: Avantor
:
- Pack type:Poly bottle
- Size:9 lbs.
- Cat. No.:JT5569-3
- Supplier no.:5569-03
- Ammonium Fluoride (NH₄F)(w/w):33.5 - 35.5 %
- Hydrofluoric Acid (HF)(w/w):7.0 - 7.3 %
- Residue after Ignition:≤10 ppm
- Trace Impurities - Aluminum (Al):≤200.0 ppb
- Arsenic and Antimony (as As):≤30.0 ppb
- Trace Impurities - Barium (Ba):≤1000.0 ppb
- Trace Impurities - Boron (B):≤200.0 ppb
- Trace Impurities - Cadmium (Cd):≤500.0 ppb
- Trace Impurities - Calcium (Ca):≤200.0 ppb
- Trace Impurities - Chromium (Cr):≤100.0 ppb
- Trace Impurities - Cobalt (Co):≤500.0 ppb
- Trace Impurities - Copper (Cu):≤100.0 ppb
- Trace Impurities - Gallium (Ga):≤500.0 ppb
- Trace Impurities - Germanium (Ge):≤500.0 ppb
- Trace Impurities - Gold (Au):≤300.0 ppb
- Heavy Metals (as Pb):≤1000.0 ppb
- Trace Impurities - Iron (Fe):≤200.0 ppb
- Trace Impurities - Lead (Pb):≤300.0 ppb
- Trace Impurities - Lithium (Li):≤1000.0 ppb
- Trace Impurities - Magnesium (Mg):≤200.0 ppb
- Trace Impurities - Manganese (Mn):≤200.0 ppb
- Trace Impurities - Nickel (Ni):≤200.0 ppb
- Trace Impurities - Potassium (K):≤300.0 ppb
- Trace Impurities - Silver (Ag):≤500.0 ppb
- Trace Impurities - Sodium (Na):≤300 ppb
- Trace Impurities - Strontium (Sr):≤1000.0 ppb
- Trace Impurities - Tin (Sn):≤300.0 ppb
- Trace Impurities - Titanium (Ti):≤300.0 ppb
- Trace Impurities - Zinc (Zn):≤300.0 ppb
- Particle Count - 1.0 µm and greater:≤25 par/ml
:
:8,II
:Management of Change (MOC) category = R
:2922
:GHS05
:GHS06
:GHS08