Sulfuric acid 50%, CMOS for the electronics industry, J.T.Baker®
Catalog # JT5253-9
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:PE drum, black
- Size:600 lbs.
- Environmentally Preferable:
- Cat. No.:JT5253-9
- Supplier no.:5253-09
Specifications
Specification Test Results
- Assay (H₂SO₄):49.0 - 51.0 %
- Color (APHA):≤10
- Residue after Ignition:≤3 ppm
- Chloride (Cl):≤1 ppm
- Nitrate (NO₃):≤0.2 ppm
- Phosphate (PO₄):≤0.5 ppm
- Trace Impurities - Aluminum (Al):≤0.100 ppm
- Arsenic and Antimony (as As):≤0.005 ppm
- Trace Impurities - Barium (Ba):≤1.000 ppm
- Trace Impurities - Boron (B):≤0.010 ppm
- Trace Impurities - Cadmium (Cd):≤0.500 ppm
- Trace Impurities - Calcium (Ca):≤1 ppm
- Trace Impurities - Chromium (Cr):≤0.500 ppm
- Trace Impurities - Cobalt (Co):≤0.500 ppm
- Trace Impurities - Copper (Cu):≤0.010 ppm
- Trace Impurities - Gallium (Ga):≤0.050 ppm
- Trace Impurities - Germanium (Ge):≤0.500 ppm
- Trace Impurities - Gold (Au):≤0.500 ppm
- Trace Impurities - Iron (Fe):≤0.2 ppm
- Trace Impurities - Lead (Pb):≤0.400 ppm
- Trace Impurities - Lithium (Li):≤1.000 ppm
- Trace Impurities - Magnesium (Mg):≤1.000 ppm
- Trace Impurities - Manganese (Mn):≤0.500 ppm
- Trace Impurities - Nickel (Ni):≤0.1 ppm
- Trace Impurities - Potassium (K):≤1.000 ppm
- Trace Impurities - Silicon (Si):≤0.500 ppm
- Trace Impurities - Silver (Ag):≤0.500 ppm
- Trace Impurities - Sodium (Na):≤1.000 ppm
- Trace Impurities - Strontium (Sr):≤0.500 ppm
- Trace Impurities - Tin (Sn):≤0.500 ppm
- Trace Impurities - Zinc (Zn):≤0.500 ppm
Specification Test Results
About this item
:
:98.08
Boiling Pt:330
EINECS:231-639-5
UN:2796
:GHS05
Storage Temperature:Ambient
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589
Melting Pt:10.38
Density:1.1…1.4 g/cm³ (20 °C)
ADR:8 II
:Management of Change (MOC) category = R