Peanut Jar, SPI Semicon
Catalog # 87003-458
Supplier: SPI Semicon
Supplier #: WC-300MM-VF
Product Details & Documents
Vacuum formed container protects 300mm (12") silicon wafers from damage during handling, testing, and storage
Manufactured from antistatic PETG. Removable snap lid. Color: clear.