Specifications
- Description:Tuning Compound
- Environmentally Preferable:
- Cat. No.:RK31001
- Supplier no.:31001
Specifications
About this item
Standard is packaged in methylene chloride at a concentration of 2500 µg/mL in a 1 mL ampoule.
- Mixture contains Decafluorotriphenylphosphine (DFTPP)
Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).