ContactUsLinkComponent
EPA method 8040 Phenol Standard No. 1, Restek
EPA method 8040 Phenol Standard No. 1, Restek
  RK31088
 :  Restek
 :  
undefined
EPA method 8040 Phenol Standard No. 1, Restek
  RK31088
 :  Restek
 :  31088
 :  
  • Description:
    EPA Method 8040 Phenol Standard No. 1
  • Cat. No.:
    RK31088
  • Supplier no.:
    31088
Standard is packaged in isopropanol at a concentration of 2000 µg/mL in a 1 mL ampoule.
  • Mixture contains 4-Chloro-3-methylphenol, 2,4-Dichlorophenol, 2-Methyl-4,6-dinitrophenol, 3-Methylphenol, 2,4,6-Trichlorophenol, 2-Nitrophenol, 4-Nitrophenol, Pentachlorophenol, and Phenol
Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).