EPA Method 8270 and CLP–Semi-Volatiles (3/90 SOW) Base/Neutrals Surrogates, NSI
Catalog # NSC376H5
Supplier: SPEX CERTIPREP LLC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Specifications
- Description:Base/Neutrals Surrogates
- Size:5mL
- Cat. No.:NSC376H5
- Supplier no.:C-376H5
Specifications
About this item
Mixture is packaged in methylene chloride at a concentration of 5,000µg/ml in an ampule
Mixture contains 2-Fluorobiphenyl, Nitrobenzene-d5, and p-Terphenyl-d14.
An A2LA traceable standard that has been second source verified in house using an A2LA verified standard.