Order Entry
United States
ContactUsLinkComponent
EPA Methods 502.1, 502.2, 524.2, 601, 624, 8010, 8021, 8240 and 8260 Gas Mixture, NSI
EPA Methods 502.1, 502.2, 524.2, 601, 624, 8010, 8021, 8240 and 8260 Gas Mixture, NSI
Catalog # NSC303
Supplier:  SPEX CERTIPREP LLC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
EPA Methods 502.1, 502.2, 524.2, 601, 624, 8010, 8021, 8240 and 8260 Gas Mixture, NSI
Catalog # NSC303
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  C-303
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Specifications

  • Description:
    EPA Methods 502.1, 502.2, 524.2, 601, 624, 8010, 8021, 8240 and 8260 Gas Mixture
  • Cat. No.:
    NSC303
  • Supplier no.:
    C-303

Specifications

About this item

Mixture is packaged in methanol at a concentration of 1000µg/mL in a 1.5mL ampule.

Mixture contains chloroethane, dichlorodifluoromethane, fluorotrichloromethane, methyl bromide, methyl chloride, and vinyl chloride.